missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dichloro-6-methylnicotinate, 97%, Thermo Scientific™

Product Code. 10021934
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10021934 25 g 25g
10332005 5 g 5g
10783483 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021934 Supplier Thermo Scientific Maybridge Supplier No. SPB04498EB
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 86129-63-7
Molecular Formula C9H9Cl2NO2
Molecular Weight (g/mol) 234.076
MDL Number MFCD00173918
InChI Key ZNFJVVLTQSOWJY-UHFFFAOYSA-N
Synonym ethyl 2,4-dichloro-6-methylnicotinate, ethyl 2,4-dichloro-6-methyl-3-pyridinecarboxylate, 2,4-dichloro-6-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2,4-dichloro-6-methyl-, ethyl ester, ethyl2,4-dichloro-6-methylnicotinate, ethyl 2,4-dichloro-6-methyl nicotinate, ethyl 2,4-dichloro-6-methylnicotinate #, 2,4-dichloro-6-methylnicotinic acid ethyl ester, ethyl 2,4-dichloro-6-methyl-pyridine-3-carboxylate, 3-pyridinecarboxylicacid,2,4-dichloro-6-methyl-,ethylester
PubChem CID 604813
IUPAC Name ethyl 2,4-dichloro-6-methylpyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=C(C=C1Cl)C)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 234.08
Percent Purity 97%
Chemical Name or Material Ethyl 2, 4-dichloro-6-methylnicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.