missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Mikromol™

Product Code. 30460116
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460116 4 x 25 mg -
30052511 25 mg 25mg
30051048 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460116 Supplier Mikromol Supplier No. MM0505.0500254AMP
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate
CAS 42019-08-9
Analyte or Component Names Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate
Shipping Condition Room Temperature
Molecular Formula C19 H19 Cl O4
Certifications/Compliance ISO 17025
Synonym Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Fenofibrate Imp. E (EP), Fenofibrate Impurity E
InChI Formula InChI=1S/C19H19ClO4/c1-4-23-18(22)19(2,3)24-16-11-7-14(8-12-16)17(21)13-5-9-15(20)10-6-13/h5-12H,4H2,1-3H3
Solution Type Neat
SMILES CCOC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2
IUPAC Name ethyl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Molecular Weight (g/mol) 346.8
Quantity 4 x 25 mg
Formula Weight 346.0972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.