missing translation for 'onlineSavingsMsg'
Learn More

ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate, Thermo Scientific™

Product Code. 10505564
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10505564 100 g 100g
10066854 25 g 25g
10565733 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10505564 Supplier Thermo Scientific Maybridge Supplier No. SB00646FA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 53266-94-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00005330
InChI Key SHQNGLYXRFCPGZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
PubChem CID 104454
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(N)=N1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 100 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Ethyl 2-(2-amino-1, 3-thiazol-4-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.