missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-2,2,2-d3 Alcohol, TRC

Product Code. 30380243
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380243 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380243 Supplier Toronto Research Chemicals Supplier No. TRCE897502500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl-2,2,2-d3 Alcohol
CAS 1759-87-1
Molecular Formula C2 D3 H3 O
Quantity 500 mg
Synonym Ethyl-2,2,2-d3 Alcohol, Ethan-2,2,2-d3-ol (9CI), Ethyl-2,2,2-d3 alcohol (6CI, 7CI, 8CI), 2,2,2-Trideuterioethanol, 2,2,2-Trideuteroethanol, Ethanol-2,2,2-d3, Ethanol-2,2,2-D3
InChI Formula InChI=1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3
SMILES [2H]C([2H])([2H])CO
IUPAC Name 2,2,2-trideuterioethanol
Molecular Weight (g/mol) 49.0869
Formula Weight 49.0607
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.