missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate, Mikromol

Product Code. 30461149
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461149 4 x 25 mg 100mg
17974266 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30461149 Supplier Mikromol Supplier No. MM1342.0300254AMP
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate
CAS 180468-42-2
Analyte or Component Names Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate
Shipping Condition Room Temperature
Molecular Formula C18 H19 N O2
Certifications/Compliance ISO 17025
Synonym (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic acid ethyl ester, 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, ethyl ester, (S)-, (S)-1-Phenyl-3,4-dihydro-1H-isoquinolinecarboxylic acid ethyl ester, Ethyl (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate, Ethyl (1S)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydro-2-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
InChI Formula InChI=1S/C18H19NO2/c1-2-21-18(20)19-13-12-14-8-6-7-11-16(14)17(19)15-9-4-3-5-10-15/h3-11,17H,2,12-13H2,1H3/t17-/m0/s1
Solution Type Neat
SMILES CCOC(=O)N1CCc2ccccc2[C@@H]1c3ccccc3
IUPAC Name ethyl (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
Molecular Weight (g/mol) 281.35
Quantity 4 x 25 mg
Formula Weight 281.1416 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.