missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (1RS)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (Ketorolac Ethyl Ester), Mikromol™
Description
Ethyl (1RS)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (Ketorolac Ethyl Ester)
Specifications
Specifications
| Chemical Name or Material | Ketorolac Ethyl Ester |
| CAS | 108061-03-6 |
| Analyte or Component Names | Ketorolac Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ketorolac Trometamol Imp. J (EP), 5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid ethyl ester, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, ethyl ester, (+/-)-, Ketorolac ethyl ester, Ketorolac Trometamol Impurity J, Ketorolac Impurity J |
| InChI Formula | InChI=1S/C17H17NO3/c1-2-21-17(20)13-10-11-18-14(13)8-9-15(18)16(19)12-6-4-3-5-7-12/h3-9,13H,2,10-11H2,1H3 |
| SMILES | CCOC(=O)C1CCn2c1ccc2C(=O)c3ccccc3 |
| IUPAC Name | ethyl 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?