missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1H-pyrazole-4-carboxylate, 98%

Product Code. 11309899
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309899 5 g 5g
11399889 1 g 1g
11319899 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309899 Supplier Thermo Scientific Alfa Aesar Supplier No. B25351.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 37622-90-5 | C6H8N2O2 | 140.142 g/mol

Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

Solubility
Miscible with acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37622-90-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00010844
InChI Key KACZQOKEFKFNDB-UHFFFAOYSA-N
PubChem CID 142179
IUPAC Name ethyl 1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=CNN=C1

Specifications

Melting Point 75°C to 80°C
Boiling Point 138°C to 140°C (3 mmHg)
Quantity 5 g
Synonym ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
Solubility Information Miscible with acetone.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Ethyl 1H-pyrazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.