missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate (Ethyl Vincaminate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate (Ethyl Vincaminate)
Specifications
Specifications
| Chemical Name or Material | Ethyl Vincaminate |
| CAS | 40163-56-2 |
| Analyte or Component Names | Ethyl Vincaminate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Vinpocetine Imp. A (EP), Vinpocetine USP Related Compound A, Vinpocetine USP RC A, Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]-naphthyridine-12-carboxylate, Ethyl Vincaminate, Vinpocetine Related Compound A, Vinpocetine Impurity A |
| InChI Formula | InChI=1S/C22H28N2O3/c1-3-21-11-7-12-23-13-10-16-15-8-5-6-9-17(15)24(18(16)19(21)23)22(26,14-21)20(25)27-4-2/h5-6,8-9,19,26H,3-4,7,10-14H2,1-2H3/t19-,21+,22+/m1/s1 |
| SMILES | CCOC(=O)[C@@]1(O)C[C@]2(CC)CCCN3CCc4c([C@H]23)n1c5ccccc45 |
| IUPAC Name | Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]-naphthyridine-12-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction