missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methylpiperidine-4-carboxylate, 96%

Product Code. 15406077
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406077 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15406077 Supplier Thermo Scientific Alfa Aesar Supplier No. H33259.ME
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24252-37-7 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24252-37-7
Molecular Formula C9H17NO2
MDL Number MFCD00130015
Quantity 500 mg
Synonym ethyl 1-methyl-4-piperidinecarboxylate, 1-methyl-piperidine-4-carboxylic acid ethyl ester, ethyl n-methyl piperidine-4-carboxylate, n-methyl ethyl isonipecotate, 4-piperidinecarboxylic acid, 1-methyl-, ethyl ester, ethyl 1-methyl piperidine-4-carboxylate, ethyl n-methylisonipecotate, 1-methyl-4-piperidinecarboxylic acid ethyl ester, 1-methylpiperidine-4-carboxylic acid ethyl ester, ethyl 1-methyl-piperidine-4-carboxylate
InChI Key JWXOOQCMGJBSML-UHFFFAOYSA-N
SMILES CCOC(=O)C1CCN(C)CC1
IUPAC Name ethyl 1-methylpiperidine-4-carboxylate
Molecular Weight (g/mol) 171.24
PubChem CID 11062856
Formula Weight 171.24
Percent Purity 96%
Chemical Name or Material Ethyl 1-methylpiperidine-4-carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.