missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-methylimidazole-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15487688
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15487688

Brand: Thermo Scientific Alfa Aesar H61185.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 1-methylimidazole-2-carboxylate is used as a synthetic intermediate for solid phase synthesis of polyamides containing imidazole.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30148-21-1
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD02683326
InChI Key NOTZYDYZBOBDFE-UHFFFAOYSA-N
Synonym ethyl 1-methyl-1h-imidazole-2-carboxylate, ethyl1-methylimidazole-2-carboxylate, 1-methyl-1h-imidazole-2-carboxylic acid ethyl ester, 1h-imidazole-2-carboxylic acid, 1-methyl-, ethyl ester, pubchem22681, ethy 1-methylimidazole-2-carboxylate, ethyl 1-methyl-imidazole-2-carboxylate, 1-methylimidazole-2-carboxylic acid ethyl ester, 1-methyl-1h-imidazole-2-carboxylic acid, ethyl ester, 1h-imidazole-2-carboxylicacid, 1-methyl-, ethyl ester
PubChem CID 4261883
IUPAC Name ethyl 1-methylimidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1C

Specifications

Quantity 25 g
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol.
Formula Weight 154.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 1-methylimidazole-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.