missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate, TRC

Product Code. 30385874
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385874 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385874 Supplier Toronto Research Chemicals Supplier No. TRCE678795100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate
CAS 28018-10-2
Molecular Formula C16 H23 N O2
Quantity 100 mg
Synonym Pethidine Hydrochloride Imp. J (EP), Ethyl 1-Ethyl-4-phenylpiperidine-4-carboxylate, Pethidine Imp. J (EP), Pethidine Hydrochloride Impurity J, Pethidine Impurity J
InChI Formula InChI=1S/C16H23NO2/c1-3-17-12-10-16(11-13-17,15(18)19-4-2)14-8-6-5-7-9-14/h5-9H,3-4,10-13H2,1-2H3
SMILES CCOC(=O)C1(CCN(CC)CC1)c2ccccc2
IUPAC Name ethyl 1-ethyl-4-phenylpiperidine-4-carboxylate
Molecular Weight (g/mol) 261.36
Formula Weight 261.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.