missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-benzylpyrrolidine-3-carboxylate, 97%

Product Code. 15442399
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15442399

Brand: Thermo Scientific Alfa Aesar H64962.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5747-92-2 | C14H19NO2 | 233.311 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5747-92-2
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
MDL Number MFCD04039852
InChI Key CYPXEPWPTXKUPL-UHFFFAOYSA-N
Synonym 1-benzyl-pyrrolidine-3-carboxylic acid ethyl ester, ethyl 1-benzyl-pyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, ethyl ester, ethyl1-benzylpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid ethyl ester, acmc-1asr8, 1-benzyl-pyrrolidine-3-carboxylicacidethylester, ksc495s0t, 1-benzyl-3-carboethoxypyrrolidine, ethyl 1-benzyl pyrrolidine-3-carboxylate
PubChem CID 11622765
IUPAC Name ethyl 1-benzylpyrrolidine-3-carboxylate
SMILES CCOC(=O)C1CCN(C1)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 25 g
Beilstein 398388
Formula Weight 233.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 1-benzylpyrrolidine-3-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.