missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10634015
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10634015 1 g Amber Glass Bottle 1g
11376835 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10634015 Supplier Thermo Scientific Maybridge Supplier No. CC59023DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-61-1
Molecular Formula C13H19N3O2
Molecular Weight (g/mol) 249.31
MDL Number MFCD09817500
InChI Key MTWGDARCNWTWJN-UHFFFAOYNA-N
Synonym ethyl 1-6-methylpyrazin-2-yl piperidine-3-carboxylate
PubChem CID 24229600
IUPAC Name ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCN(C1)C1=NC(C)=CN=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material Ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.