missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(4-cyanopyrid-2-yl)piperidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. 11310687
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11310687 250 mg Amber Glass Bottle 250mg
10769035 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310687 Supplier Thermo Scientific Maybridge Supplier No. CC50716CB
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-67-8
Molecular Formula C14H17N3O2
Molecular Weight (g/mol) 259.31
MDL Number MFCD09025864
InChI Key IAIJZMCYSMAKQF-UHFFFAOYSA-N
Synonym ethyl 1-4-cyanopyridin-2-yl piperidine-4-carboxylate, ethyl 1-4-cyanopyrid-2-yl piperidine-4-carboxylate, ethyl 1-4-cyano-2-pyridyl piperidine-4-carboxylate
PubChem CID 18525853
IUPAC Name ethyl 1-(4-cyanopyridin-2-yl)piperidine-4-carboxylate
SMILES CCOC(=O)C1CCN(CC1)C1=NC=CC(=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 259.31
Percent Purity 97%
Chemical Name or Material Ethyl 1-(4-cyanopyrid-2-yl)piperidine-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.