missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(4-cyanopyrid-2-yl)piperidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10769035
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-67-8
Molecular Formula C14H17N3O2
Molecular Weight (g/mol) 259.31
MDL Number MFCD09025864
InChI Key IAIJZMCYSMAKQF-UHFFFAOYSA-N
Synonym ethyl 1-4-cyanopyridin-2-yl piperidine-4-carboxylate, ethyl 1-4-cyanopyrid-2-yl piperidine-4-carboxylate, ethyl 1-4-cyano-2-pyridyl piperidine-4-carboxylate
PubChem CID 18525853
IUPAC Name ethyl 1-(4-cyanopyridin-2-yl)piperidine-4-carboxylate
SMILES CCOC(=O)C1CCN(CC1)C1=NC=CC(=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 259.31
Percent Purity 97%
Chemical Name or Material Ethyl 1-(4-cyanopyrid-2-yl)piperidine-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'