missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,3-dithiolane-2-carboxylate, 99%, Thermo Scientific™

Product Code. 10358010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10358010 25 mL Glass bottle 25mL
10348010 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10358010 Supplier Thermo Scientific Acros Supplier No. 209990250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20461-99-8
Molecular Formula C6H10O2S2
Molecular Weight (g/mol) 178.26
MDL Number MFCD00005411
InChI Key OMCSHTHLIQOHDD-UHFFFAOYSA-N
Synonym 2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
PubChem CID 88551
IUPAC Name ethyl 1,3-dithiolane-2-carboxylate
SMILES CCOC(=O)C1SCCS1

Specifications

Color Yellow
Density 1.2400g/mL
Boiling Point 85°C (0.1 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5380 to 1.54
Quantity 25 mL
Beilstein 19, II, 290
Specific Gravity 1.24
Formula Weight 178.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 1, 3-dithiolane-2-carboxylate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.