missing translation for 'onlineSavingsMsg'
Learn More

Ethoxytrimethylsilane, 95%

Product Code. 10307911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307911 25 g Glass bottle 25g
10469481 5 g Glass bottle 5g
10016293 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307911 Supplier Thermo Scientific Supplier No. 313820250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1825-62-3 | C5H14OSi | 118.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1825-62-3
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00009069
InChI Key RSIHJDGMBDPTIM-UHFFFAOYSA-N
Synonym trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
PubChem CID 15772
IUPAC Name ethoxy(trimethyl)silane
SMILES CCO[Si](C)(C)C

Specifications

Color Colorless
Density 0.7500g/mL
Boiling Point 75°C to 76°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Si(CH3)3OC2H5
Refractive Index 1.3730 to 1.375
Quantity 25 g
Specific Gravity 0.75
Solubility Information Solubility in water: reacts with water
Formula Weight 118.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethoxytrimethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethoxytrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethoxytrimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.