missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyquin, Honeywell Fluka™

Product Code. 16604484 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16604484

Brand: Honeywell Fluka R1749100MG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethoxyquin
CAS 91-53-2
CAS Max % ≤ 100%
Density 1.031 g/cm3 (25°C)
Flash Point 150°C
Beilstein 158223
Color Yellow
Melting Point <-20°C
Boiling Point 123°C to 125°C (1013 hPa)
Synonym 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline; 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline
Molecular Weight (g/mol) 217.3 g/mol
Quantity 100 mg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.