missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxyoxalyl Chloride, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Ethoxyoxalyl Chloride |
| CAS | 4755-77-5 |
| Molecular Formula | C4H5ClO3 |
| Quantity | 50 g |
| Synonym | Chlorooxoacetic Acid Ethyl Ester, Chloroglyoxylic Acid Ethyl Ester, 1-(Chlorocarbonyl)formic Acid Ethyl Ester, Ethoxalyl Chloride, Ethyl (Chlorocarbonyl)carboxylate, Ethyl (Chloroformyl)formate, Ethyl 2-Chloro-2-oxoacetate, Ethyl 2-Chloro-2-oxoethanoate, Ethyl Chloroglyoxylate, Ethyl Chlorooxalate, Ethyl Chlorooxoacetate, Ethyl Oxaloyl Chloride, Ethyl Oxalyl Chloride, Monoethyl Oxalate Chloride, Monoethyl Oxalate Monochloride, Monoethyl Oxaloyl Chloride, NSC 80644, Oxalic Acid Chloride Ethyl Ester, Oxalic Acid Ethyl Ester Chloride, Oxalic Acid Monoethyl Ester Chloride, Oxalic Acid Monoethyl Ester Monochloride, Oxalyl Chloride Ethyl Ester |
| InChI Formula | InChI=1S/C4H5ClO3/c1-2-8-4(7)3(5)6/h2H2,1H3 |
| SMILES | CCOC(=O)C(=O)Cl |
| IUPAC Name | ethyl 2-chloro-2-oxoacetate |
| Molecular Weight (g/mol) | 136.53 |
| Formula Weight | 135.9927 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction