missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyoxalyl Chloride, TRC

Product Code. 30370264
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30370264

Brand: Toronto Research Chemicals TRCE89246025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethoxyoxalyl Chloride
CAS 4755-77-5
Molecular Formula C4H5ClO3
Quantity 25 g
Synonym Chlorooxoacetic Acid Ethyl Ester, Chloroglyoxylic Acid Ethyl Ester, 1-(Chlorocarbonyl)formic Acid Ethyl Ester, Ethoxalyl Chloride, Ethyl (Chlorocarbonyl)carboxylate, Ethyl (Chloroformyl)formate, Ethyl 2-Chloro-2-oxoacetate, Ethyl 2-Chloro-2-oxoethanoate, Ethyl Chloroglyoxylate, Ethyl Chlorooxalate, Ethyl Chlorooxoacetate, Ethyl Oxaloyl Chloride, Ethyl Oxalyl Chloride, Monoethyl Oxalate Chloride, Monoethyl Oxalate Monochloride, Monoethyl Oxaloyl Chloride, NSC 80644, Oxalic Acid Chloride Ethyl Ester, Oxalic Acid Ethyl Ester Chloride, Oxalic Acid Monoethyl Ester Chloride, Oxalic Acid Monoethyl Ester Monochloride, Oxalyl Chloride Ethyl Ester
InChI Formula InChI=1S/C4H5ClO3/c1-2-8-4(7)3(5)6/h2H2,1H3
SMILES CCOC(=O)C(=O)Cl
IUPAC Name ethyl 2-chloro-2-oxoacetate
Molecular Weight (g/mol) 136.53
Formula Weight 135.9927
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.