missing translation for 'onlineSavingsMsg'
Learn More

Ethoxymethylenemalononitrile, 98%

Product Code. p-3778527 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10031050

Brand: Thermo Scientific Acros 117921000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-06-8 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-06-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00001854
InChI Key OEICGMPRFOJHKO-UHFFFAOYSA-N
Synonym ethoxymethylenemalononitrile, ethoxymethylene malononitrile, propanedinitrile, ethoxymethylene, 2-ethoxymethylene malononitrile, 2-ethoxymethylidene propanedinitrile, 2-cyano-3-ethoxyacrylonitrile, ethoxymethylene propanedinitrile, usaf a-9230, usaf kf-10
PubChem CID 67152
IUPAC Name 2-(ethoxymethylidene)propanedinitrile
SMILES CCOC=C(C#N)C#N

Specifications

Melting Point 63°C to 68°C
Color Colorless to Yellow
Density 155g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 98%
Linear Formula C2H5OCH=C(CN)2
Beilstein 03, I, 162
Formula Weight 122.13
Percent Purity ≥97.5% (GC)
Chemical Name or Material Ethoxymethylenemalononitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.