missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethylene)triphenylphosphorane, 98+%

Product Code. 11455343
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455343 10 g 10g
11465343 50 g 50g
11475343 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455343 Supplier Thermo Scientific Alfa Aesar Supplier No. A12896.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1099-45-2 | C22H21O2P | 348.38 g/mol

(Ethoxycarbonylmethylene)triphenylphosphoran is used as a Wittig reagent in organic synthesis. It is used in the preparation of indole from o-nitrobenzaldehydes, coumarins from o-hydroxy acetophenone. It acts as an inhibitor and inhibits the activity of cholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Ethoxycarbonylmethylene)triphenylphosphoran is used as a Wittig reagent in organic synthesis. It is used in the preparation of indole from o-nitrobenzaldehydes, coumarins from o-hydroxy acetophenone. It acts as an inhibitor and inhibits the activity of cholinesterase.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1099-45-2
Molecular Formula C22H21O2P
Molecular Weight (g/mol) 348.38
MDL Number MFCD00009183
InChI Key IIHPVYJPDKJYOU-UHFFFAOYSA-N
Synonym carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
PubChem CID 70670
IUPAC Name ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 126°C to 130°C
Quantity 10 g
Beilstein 754639
Sensitivity Air sensitive
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 348.38
Percent Purity ≥98%
Chemical Name or Material (Ethoxycarbonylmethylene)triphenylphosphorane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.