missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)triphenylphosphonium-13C2 Bromide, TRC

Product Code. 30375043
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375043 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375043 Supplier Toronto Research Chemicals Supplier No. TRCE891452250MG
 Restricted Item
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Ethoxycarbonylmethyl)triphenylphosphonium-13C2 Bromide
CAS 109376-35-4
Molecular Formula C2013C2H22BrO2P
Quantity 250 mg
Synonym (2-Ethoxy-2-oxoethyl)(triphenyl)phosphonium-13C2 Bromide, (Carbethoxymethyl)triphenylphosphonium-13C2 Bromide, Carboethoxymethyltriphenylphosphonium-13C2 Bromide, Ethoxycarbonylmethyltriphenylphosphonium-13C2 Bromide, NSC 60450-13C2
InChI Formula InChI=1S/C22H22O2P.BrH/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/i18+1,22+1;
SMILES OC(=O)c1c(C)onc1
Molecular Weight (g/mol) 431.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.