missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isothiocyanate, 97%

Product Code. 11394354
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394354 25 g 25g
11384354 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394354 Supplier Thermo Scientific Alfa Aesar Supplier No. L00911.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16182-04-0 | C4H5NO2S | 131.149 g/mol

Ethoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform and ether.

Notes
Moisture, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16182-04-0
Molecular Formula C4H5NO2S
Molecular Weight (g/mol) 131.149
MDL Number MFCD00004814
InChI Key BDTDECDAHYOJRO-UHFFFAOYSA-N
Synonym ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate
PubChem CID 85320
IUPAC Name ethyl N-(sulfanylidenemethylidene)carbamate
SMILES CCOC(=O)N=C=S

Specifications

Density 1.11
Boiling Point 44°C to 46°C (10 mmHg)
Flash Point 50°C (122°F)
Odor Pungent
Refractive Index 1.503
Quantity 25 g
UN Number UN2924
Beilstein 606091
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and ether.
Formula Weight 131.15
Percent Purity 97%
Chemical Name or Material Ethoxycarbonyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.