missing translation for 'onlineSavingsMsg'
Learn More

Ethoxycarbonyl isocyanate, 90%, tech.

Product Code. 10412845 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412845 5 g Glass bottle 5g
10561151 1 g Glass bottle 1g
11945681 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10412845 Supplier Thermo Scientific Acros Supplier No. 314310050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19617-43-7 | C4H5NO3 | 115.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19617-43-7
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
InChI Key VMVZGGPZNHFGKS-UHFFFAOYSA-N
Synonym ethoxycarbonyl isocyanate, ethyl isocyanatoformate, ethyl carbonisocyanatidate, ethyl n-carbonylcarbamate, ethoxycarbonylisocyanate, carbonisocyanatidic acid, ethyl ester, ethyl isocyanatooate, ethylisocyanatoformate, acmc-20al2o, carbonisocyanatidicacid,ethylester
PubChem CID 300166
IUPAC Name ethyl N-(oxomethylidene)carbamate
SMILES CCOC(=O)N=C=O

Specifications

Color Colorless
Density 1.1100g/mL
Boiling Point 25.0°C (10.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CNCO
Refractive Index 1.4060 to 1.4090
Quantity 5 g
Specific Gravity 1.11
Formula Weight 115.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethoxycarbonyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.