missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyamine Hydrochloride, TRC

Product Code. 30382612
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382612 500 mg -
30382006 100 mg -
30379736 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382612 Supplier Toronto Research Chemicals Supplier No. TRCE676320500MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3332-29-4
Molecular Formula C2H7NO . HCl
Molecular Weight (g/mol) 97.54
Synonym Ethoxylamine Hydrochloride, N-Ethoxyamine Hydrochloride, O-Ethylhydroxylamine Hydrochloride, (Aminooxy)ethane Hydrochloride, Ethoxamine Hydrochloride, Ethoxyammonium Chloride
IUPAC Name O-ethylhydroxylamine;hydrochloride
SMILES Cl.CCON

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2H7NO.ClH/c1-2-4-3;/h2-3H2,1H3;1H
Formula Weight 97.0294
Chemical Name or Material Ethoxyamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.