missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic Acid, TRC

Product Code. 30373731
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373731 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373731 Supplier Toronto Research Chemicals Supplier No. TRCE8905985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 627-03-2
Molecular Formula C4H8O3
Quantity 5 g
InChI Formula InChI=1S/C4H8O3/c1-2-7-3-4(5)6/h2-3H2,1H3,(H,5,6)
SMILES CCOCC(=O)O
Molecular Weight (g/mol) 104.11
Formula Weight 104.0473
Chemical Name or Material Ethoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.