missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic acid, 98%

Product Code. 10774661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10774661 25 g Glass bottle 25g
10103070 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10774661 Supplier Thermo Scientific Acros Supplier No. 159860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-03-2 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-03-2
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004310
InChI Key YZGQDNOIGFBYKF-UHFFFAOYSA-N
Synonym ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
PubChem CID 12301
IUPAC Name 2-ethoxyacetic acid
SMILES CCOCC(=O)O

Specifications

Color Colorless to Yellow
Density 1.1020g/mL
Boiling Point 97°C to 100°C (11.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5OCH2CO2H
Refractive Index 1.4170 to 1.42
Quantity 25 g
Beilstein 03, 233
Specific Gravity 1.102
Solubility Information Solubility in water: soluble in water
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethoxyacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Ethoxyacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.