missing translation for 'onlineSavingsMsg'
Learn More

Ethotoin-d5, TRC

Product Code. 30386363
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386363 50 mg
30381728 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386363 Supplier Toronto Research Chemicals Supplier No. TRCE89053750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H7D5N2O2
Molecular Weight (g/mol) 209.26
Synonym (±)-Ethotoin-d5, 1-Ethyl-2,5-dioxo-4-phenylimidazolidine-d5, 3-Ethyl-5-phenyl-2,4-imidazolidinedione-d5, 3-Ethyl-5-phenylhydantoin-d5, 3-Ethyl-5-phenylimidazolidin-2,4-dione-d5, AC-695, Accenon, Peganone-d5, 3-Ethyl-5-phenyl-2,4-Imidazolidinedione-d5
IUPAC Name 3-(ethyl-d5)-5-phenylimidazolidine-2,4-dione
SMILES O=C(NC1C2=CC=CC=C2)N(C([2H])([2H])C([2H])([2H])[2H])C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H12N2O2/c1-2-13-10(14)9(12-11(13)15)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,12,15)/i1D3,2D2
Formula Weight 209.13
Chemical Name or Material Ethotoin-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.