missing translation for 'onlineSavingsMsg'
Learn More

Ethosuximide

Product Code. 15666804 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15666804 25 g 25g
15656804 5 g 5g
15648735 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15666804 Supplier Thermo Scientific Acros Supplier No. 461400250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-67-8 | C7H11NO2 | 141.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-67-8
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00072123
InChI Key HAPOVYFOVVWLRS-UHFFFAOYNA-N
Synonym ethosuximide, zarontin, etosuximida, 2-ethyl-2-methylsuccinimide, ethosuxide, ethosuccimide, ethosuccinimide, petnidan, suxinutin, atysmal
PubChem CID 3291
ChEBI CHEBI:4887
IUPAC Name 3-ethyl-3-methylpyrrolidine-2,5-dione
SMILES CCC1(C)CC(=O)NC1=O

Specifications

Melting Point 47.0°C to 50.0°C
Boiling Point 134.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 141.17
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Ethosuximide
compliance-icons compliance-icons
Product Identifier
  • Ethosuximide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H341-Suspected of causing genetic defects.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethosuximide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.