missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethosuximide
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 47.0°C to 50.0°C |
| Boiling Point | 134.0°C (5.0 mmHg) |
| Infrared Spectrum | Authentic |
| Quantity | 100 g |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 141.17 |
| Percent Purity | ≥97.5% (HPLC) |
| Chemical Name or Material | Ethosuximide |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu