missing translation for 'onlineSavingsMsg'
Learn More

Ethosuximide, MedChemExpress

Product Code. 16402276
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402276 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402276 Supplier Medchem Express Supplier No. HYB1378
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ethosuximide, a widely prescribed anti-epileptic drug, improves the phenotypes of multiple neurodegenerative disease models and blocks the low voltage activated T-type calcium channel.

Ethosuximide, a widely prescribed anti-epileptic drug, improves the phenotypes of multiple neurodegenerative disease models and blocks the low voltage activated T-type calcium channel.

Specifications

Chemical Name or Material Ethosuximide
CAS 77-67-8
Color White
Purity Grade Notes Research
Molecular Formula C7H11NO2
Quantity 500 mg
Solubility Information DMSO : 100 mg/mL (708.37 mM; Need ultrasonic)
SMILES O=C(C(C)(CC)C1)NC1=O
Molecular Weight (g/mol) 141.17
Formula Weight 141.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.