missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoprophos (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Ethoprophos |
| CAS | 13194-48-4 |
| Molecular Formula | C8 H19 O2 P S2 |
| Quantity | 10 g |
| Synonym | Phosphorodithioic acid, O-ethyl S,S-dipropyl ester, Ethyl propyl phosphorodithioate ((EtO)(PrS)2PO) (7CI), ENT 27318, Ethoprop, Ethoprophos, Mobile, Mocap, Mocap 10G, Mocap 15G, Mocap 6EC, O-Ethyl S,S-dipropyl dithiophosphate, O-Ethyl S,S-dipropyl phosphorodithioate, Profos, Prophos, Prophos (ester), Rovokil, S,S-Dipropyl O-ethyl phosphorodithioate, VC 9-104 |
| InChI Formula | InChI=1S/C8H19O2PS2/c1-4-7-12-11(9,10-6-3)13-8-5-2/h4-8H2,1-3H3 |
| SMILES | CCCSP(=O)(OCC)SCCC |
| IUPAC Name | 1-[ethoxy(propylsulfanyl)phosphoryl]sulfanylpropane |
| Molecular Weight (g/mol) | 242.34 |
| Formula Weight | 242.0564 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?