missing translation for 'onlineSavingsMsg'
Learn More

Ethopabate, TRC

Product Code. 30135382
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135382 2.5 g 2.5g
30010304 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135382 Supplier Toronto Research Chemicals Supplier No. TRCE8908002.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-06-3
Molecular Formula C12 H15 N O4
Molecular Weight (g/mol) 237.25
Synonym Benzoic acid, 4-(acetylamino)-2-ethoxy-, methyl ester, Benzoic acid, 4-acetamido-2-ethoxy-, methyl ester (7CI,8CI), 4-Acetamido-2-ethoxybenzoic acid methyl ester, Ethopabate, Ethyl pabate, Methyl 2-ethoxy-4-acetamidobenzoate, Methyl 4-(acetylamino)-2-ethoxybenzoate, Methyl 4-acetamido-2-ethoxybenzoate
IUPAC Name methyl 4-acetamido-2-ethoxybenzoate
SMILES CCOc1cc(NC(=O)C)ccc1C(=O)OC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H15NO4/c1-4-17-11-7-9(13-8(2)14)5-6-10(11)12(15)16-3/h5-7H,4H2,1-3H3,(H,13,14)
Formula Weight 237.1001
Chemical Name or Material Ethopabate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.