missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethisterone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Ethisterone
Specifications
Specifications
| Chemical Name or Material | Ethisterone |
| CAS | 434-03-7 |
| Analyte or Component Names | Ethisterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Testosterone, 17-ethynyl- (5CI), 17-Ethynyl-17-hydroxy-3-ketoandrostene-4, Aethisteron, Anhydrohydroxyprogesterone, Anhydroxyprogesterone, Colutoid, Ethinone, Ethinyltestosterone, Ethisterone, Ethynyltestosterone, Gestoral, Lucorteum Oral, Lutidon Oral, Lutocyclin-Linguetten, Lutocyclol, Lutocylol, NSC 9565, Nalutoral, Nugestoral, Ora-Lutin, Pranone, Pregneninolone, Pregnin, Primolut C, Prodoxan, Prodroxan, Produxan, Progestab, Progestin P, Progestolets, Progestoral, Prolutol, Proluton C, Prone, Syngestrotabs, Trosinone, Pregn-4-en-20-yn-3-one, 17-hydroxy-, (17alpha)-, 17alpha-Pregn-4-en-20-yn-3-one, 17-hydroxy- (6CI,8CI), 17-Ethynyl-17beta-hydroxyandrost-4-en-3-one, 17-Hydroxy-17alpha-pregn-4-en-20-yn-3-one, 17alpha-Ethynyl-17-hydroxyandrost-4-en-3-one, 17alpha-Ethynyl-17beta-hydroxy-4-androsten-3-one, 17alpha-Ethynyl-4-androsten-17beta-ol-3-one, 17alpha-Ethynyltestosterone, 17beta-Hydroxy-17alpha-ethynylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-pregn-4-en-20-yn-3-one, 4-Androsten-17alpha-ethinyl-17beta-ol-3-one, Pregn-4-en-17beta-ol-3-on-20-yne |
| InChI Formula | InChI=1S/C21H28O2/c1-4-21(23)12-9-18-16-6-5-14-13-15(22)7-10-19(14,2)17(16)8-11-20(18,21)3/h1,13,16-18,23H,5-12H2,2-3H3/t16-,17+,18+,19+,20+,21+/m1/s1 |
| SMILES | C[C@]12CCC(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]4(O)C#C |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction