missing translation for 'onlineSavingsMsg'
Learn More

Ethisterone, 98%, Thermo Scientific™

£139.99 - £306.00

Chemical Identifiers

CAS 434-03-7
Molecular Formula C21H28O2
Molecular Weight (g/mol) 312.453
MDL Number MFCD00003656
InChI Key CHNXZKVNWQUJIB-SJFWLOONSA-N
Synonym pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one
PubChem CID 12016247
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435779
SDS View Documents
Thermo Scientific Alfa Aesar
J63580.06
5g
ONLINE EXCLUSIVE PRICE
£143.00
£139.99 / 5g
Save £3.01
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15445779
SDS View Documents
Thermo Scientific Alfa Aesar
J63580.14
25g
ONLINE EXCLUSIVE PRICE
£321.00
£306.00 / 25g
Save £15.00
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
soluble in chloroform, DMSO (0.05 mg/ml), acetone (slightly ), ethanol (<1mg/mL at 25°C), and water (<1mg/mL at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

434-03-7
312.453
CHNXZKVNWQUJIB-SJFWLOONSA-N
12016247
CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C
C21H28O2
MFCD00003656
pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one
(8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one

Specifications

434-03-7
MFCD00003656
14,3741
soluble in chloroform,DMSO (0.05mg/ml),acetone (slightly ),ethanol (<1mg/ml at 25°C),and water (<1mg/ml at 25°C).
CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C
312.453
312.45
Ethisterone
C21H28O2
1889895
pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one
CHNXZKVNWQUJIB-SJFWLOONSA-N
(8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
12016247
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H351-H360-H362

missing translation for 'einecsNumber' : 207-096-5

missing translation for 'rtecsNumber' : TU5570250

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.