missing translation for 'onlineSavingsMsg'
Learn More

Ethirimol, TRC

Product Code. 30377015
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377015 10 g
30386073 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377015 Supplier Toronto Research Chemicals Supplier No. TRCE89020010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23947-60-6
Molecular Formula C11 H19 N3 O
Molecular Weight (g/mol) 209.29
Synonym 4(1H)-Pyrimidinone, 5-butyl-2-(ethylamino)-6-methyl- (9CI), 5-Butyl-2-(ethylamino)-6-methyl-4(3H)-pyrimidinone, 5-Butyl-2-ethylamino-6-hydroxy-4-methylpyrimidine, ETH 560, Ethirimol, Milgo, Milgo E, Milstem, Milstem Seed Dressing, NSC, 4(3H)-Pyrimidinone, 5-butyl-2-(ethylamino)-6-methyl-263491, PP 149
IUPAC Name 5-butyl-2-(ethylamino)-6-methylpyrimidin-4-ol
SMILES CCCCc1c(C)nc(NCC)nc1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H19N3O/c1-4-6-7-9-8(3)13-11(12-5-2)14-10(9)15/h4-7H2,1-3H3,(H2,12,13,14,15)
Formula Weight 209.1528
Chemical Name or Material Ethirimol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.