missing translation for 'onlineSavingsMsg'
Learn More

Ethiprole Amide, TRC

Product Code. 30381141
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381141 250 mg 250mg
30372127 50 mg 50mg
30385161 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381141 Supplier Toronto Research Chemicals Supplier No. TRCE623150250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623151-90-6
Molecular Formula C13H11Cl2F3N4O2S
Molecular Weight (g/mol) 415.22
Synonym 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(ethylsulfinyl)-1H-pyrazole-3-carboxamide
SMILES CCS(=O)c1c(N)n(nc1C(=O)N)c2c(Cl)cc(cc2Cl)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11Cl2F3N4O2S/c1-2-25(24)10-8(12(20)23)21-22(11(10)19)9-6(14)3-5(4-7(9)15)13(16,17)18/h3-4H,2,19H2,1H3,(H2,20,23)
Formula Weight 413.9932
Chemical Name or Material Ethiprole Amide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.