missing translation for 'onlineSavingsMsg'
Learn More

Ethion, TRC

Product Code. 30377554
Change view
Click to view available options
Quantity:
1 g
10 g
Product Code. Quantity
30377554 1 g
30379835 10 g
2 options
This item is not returnable. View return policy

Product Code. 30377554

Brand: Toronto Research Chemicals TRCE6783051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-12-2
Formula Weight 383.9876
InChI Formula InChI=1S/C9H22O4P2S4/c1-5-10-14(16,11-6-2)18-9-19-15(17,12-7-3)13-8-4/h5-9H2,1-4H3
IUPAC Name diethoxyphosphinothioylsulfanylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Formula C9 H22 O4 P2 S4
Molecular Weight (g/mol) 384.48
SMILES CCOP(=S)(OCC)SCSP(=S)(OCC)OCC
Synonym Phosphorodithioic acid, S,S'-methylene O,O,O',O'-tetraethyl ester (8CI,9CI), Diethion, ENT 24,105, Ethiol 100, Ethion, Ethopaz, Ethyl methylene phosphorodithioate ([(EtO)2P(S)S]2CH2), Fosfatox E, Hylemax, Hylemox, Match, Niagara 1240, Nialate, O,O,O',O'-Tetraethyl S,S'-methylene bis(phosphorodithioate), Phosphotox E, RP 8167, Rhodocide, Rodocid, Rodocide, Tetraethyl S,S'-methylene bis(phosphorothiolothionate), Tomahawk
Chemical Name or Material Ethion

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.