missing translation for 'onlineSavingsMsg'
Learn More

Ethion Dioxon, TRC

Product Code. 30386470
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386470 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386470 Supplier Toronto Research Chemicals Supplier No. TRCE28468025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethion-dioxon
CAS 22756-17-8
Purity Grade Notes GC
Molecular Formula C9 H22 O6 P2 S2
Quantity 25 mg
Synonym Ethion Dioxon, Ethyl methylene phosphorothioate ((EtO)4(CH2S2)P2O2) (6CI), Ethion dioxon, Ethion oxygen analog, Methanedithiol, S,S-diester with O,O-diethyl phosphorothioate, PO-Ethion, Ethion-dioxon
InChI Formula InChI=1S/C9H22O6P2S2/c1-5-12-16(10,13-6-2)18-9-19-17(11,14-7-3)15-8-4/h5-9H2,1-4H3
SMILES CCOP(=O)(OCC)SCSP(=O)(OCC)OCC
IUPAC Name 1-[diethoxyphosphorylsulfanylmethylsulfanyl(ethoxy)phosphoryl]oxyethane
Molecular Weight (g/mol) 352.345
Formula Weight 352.033
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.