missing translation for 'onlineSavingsMsg'
Learn More

Ethinylestradiol, 98%, Thermo Scientific Chemicals

Product Code. 15660984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15660984 1 g 1g
15648745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15660984 Supplier Thermo Scientific Acros Supplier No. 461550010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-63-6
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00003690
InChI Key BFPYWIDHMRZLRN-SLHNCBLASA-N
Synonym ethinyl estradiol, ethynylestradiol, ethynyl estradiol, ethinylestradiol, ethinyloestradiol, ginestrene, 17-ethinylestradiol, progynon c, ethinoral, eticyclin
PubChem CID 5991
ChEBI CHEBI:4903
IUPAC Name (8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 180.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 296.41
Percent Purity 98%
Chemical Name or Material Ethinylestradiol
compliance-icons compliance-icons
Product Identifier
  • Ethinylestradiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethinylestradiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.