missing translation for 'onlineSavingsMsg'
Learn More

Ethidium Bromide, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15518204 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 g
Packaging:
POLYSTORMOR
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15518204

Brand: Avantor J.T.Baker 400700

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Nucleic Acid Detection in Electrophoresis

Specifications

CAS 1239-45-8
Packaging POLYSTORMOR
Molecular Formula C21H20BrN3
MDL Number MFCD00011724
Quantity 1 g
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Molecular Weight (g/mol) 394.32
PubChem CID 14710
ChEBI CHEBI:4883
Formula Weight 394.31
Grade Ultrapure Bioreagent
Chemical Name or Material Ethidium Bromide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.