missing translation for 'onlineSavingsMsg'
Learn More

Ethidium Bromide, TRC

Product Code. 30380688
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30380688

Brand: Toronto Research Chemicals TRCE8907005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethidium Bromide
CAS 1239-45-8
Molecular Formula C21H20BrN3
Quantity 5 g
Synonym 3,8-Diamino-5-ethyl-6-phenylphenanthridinium Bromide, 2,7-Diamino-10-ethyl-9-phenylphenanthridinium Bromide, 2,7-Diamino-9-phenyl-10-ethylphenanthridinium Bromide, 2,7-Diamino-9-phenylphenanthridine Ethobromide, Dromilac, Homidium Bromide
InChI Formula InChI=1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H
SMILES [Br-].CC[n+]1c(c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Molecular Weight (g/mol) 394.31
Formula Weight 393.0841
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.