missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethidium Bromide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Ethidium Bromide |
| CAS | 1239-45-8 |
| Molecular Formula | C21H20BrN3 |
| Quantity | 25 g |
| Synonym | 3,8-Diamino-5-ethyl-6-phenylphenanthridinium Bromide, 2,7-Diamino-10-ethyl-9-phenylphenanthridinium Bromide, 2,7-Diamino-9-phenyl-10-ethylphenanthridinium Bromide, 2,7-Diamino-9-phenylphenanthridine Ethobromide, Dromilac, Homidium Bromide |
| InChI Formula | InChI=1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H |
| SMILES | [Br-].CC[n+]1c(c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14 |
| IUPAC Name | 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide |
| Molecular Weight (g/mol) | 394.31 |
| Formula Weight | 393.0841 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction