missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
- Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
- Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.
- A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
- Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
- It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.
- MP's EtBr confers a deep redstain to DNA by acting as an intercalating agent between the starch bases.
- Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase.
Tekniske data
Tekniske data
| CAS | 1239-45-8 |
| Detection | 15-30 min with 0.5mg/mL solution |
| Color | Dark Red/Purple |
| Concentration | 10 mg/mL |
| Form | Liquid |
| For Use With (Application) | Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acidgel Electrophoresis & Blotting |
| Molecular Weight (g/mol) | 394.3 |
| Quantity | 6 x 10 mL |
| Storage Requirements | Store at +4°C, protect from light. |
| Product Type | Fluorescent DNA/RNA Stain |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?