missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mL
6 x 10 mL
Unit Size:
10mL
60mL
Description
- Concentration : 0.95 – 1.05%
- Purity (on dry basis) : > 98.0%
- RNase, DNase, Protease free
- Fluorescent DNA and RNA intercalating agent.
- Used as a dye for DNA and RNA electrophoresis.
- Storage: Store at + 4°C protected from light.
- A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
- Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.
- It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded D
- Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
Specifications
Specifications
| CAS | 1239-45-8 |
| Detection | 15-30 min with 0.5mg/mL solution |
| Color | Dark Red/Purple |
| Concentration | 10 mg/mL |
| Product Type | Fluorescent DNA/RNA Stain |
| Form | Liquid |
| For Use With (Application) | Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acidgel Electrophoresis & Blotting |
| Molecular Weight (g/mol) | 394.3 |
| Quantity | 10 mL |
| Storage Requirements | Store at +4°C, protect from light. |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- P102-Keep out of reach of children.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethidium bromide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction