Learn More
Ethidium bromide, 95%, Thermo Scientific Chemicals
£29.90 - £112.00
Chemical Identifiers
CAS | 1239-45-8 |
---|---|
Molecular Formula | C21H20BrN3 |
Molecular Weight (g/mol) | 394.32 |
MDL Number | MFCD00011724 |
InChI Key | ZMMJGEGLRURXTF-UHFFFAOYSA-N |
Synonym | ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l |
PubChem CID | 14710 |
ChEBI | CHEBI:4883 |
SMILES | [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11406750
|
Thermo Scientific Alfa Aesar
L07482.03 |
1 g |
£29.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416750
|
Thermo Scientific Alfa Aesar
L07482.06 |
5 g |
£112.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For DNA isolation and visualization
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1239-45-8 | |
394.32 | |
ZMMJGEGLRURXTF-UHFFFAOYSA-N | |
14710 | |
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12 |
C21H20BrN3 | |
MFCD00011724 | |
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l | |
CHEBI:4883 |
Specifications
1239-45-8 | |
Weak | |
MFCD00011724 | |
3642536 | |
ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l | |
[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12 | |
394.32 | |
CHEBI:4883 | |
98% | |
Ethidium bromide |
∽261°C (decomposition) | |
C21H20BrN3 | |
UN2811 | |
14,4731 | |
ZMMJGEGLRURXTF-UHFFFAOYSA-N | |
5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide | |
14710 | |
394.32 | |
(dry wt.) |
Safety and Handling
GHS H Statement
H330-H341-H302
Fatal if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P304+P340-P308+P313-P310-P330-P501c
H302-H330-H341
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 214-984-6
missing translation for 'rtecsNumber' : SF7950000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only