missing translation for 'onlineSavingsMsg'
Learn More

Ether, Ultra Low Water, Contains No Preservative, BakerDRY™, J.T. Baker™

Catalog Number p-5391130
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Flash Point −45.00°C
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Quantity 1 L
Specific Gravity 1L = 0.71kg
Formula Weight 74.12
Grade BakerDRY™
Chemical Name or Material Ether, Ultra Low Water, Contains No Preservative
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'