missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethephon, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Ethephon |
| CAS | 16672-87-0 |
| Analyte or Component Names | Ethephon |
| Shipping Condition | +20°C |
| Molecular Formula | C2 H6 Cl O3 P |
| Certifications/Compliance | ISO/IEC 17025 |
| InChI Formula | InChI=1S/C2H6ClO3P/c3-1-2-7(4,5)6/h1-2H2,(H2,4,5,6) |
| SMILES | OP(=O)(O)CCCl |
| IUPAC Name | 2-chloroethylphosphonic acid |
| Molecular Weight (g/mol) | 144.49 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction