missing translation for 'onlineSavingsMsg'
Learn More

Ethenoadenosine Antibody (1G4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18424694 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424694 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424694 Supplier Novus Biologicals Supplier No. NB600442JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ethenoadenosine Monoclonal antibody specifically detects Ethenoadenosine in All samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ethenoadenosine
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 1G4
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C12H13N5O4, N6-Ethenoadenosine
Host Species Mouse
Immunogen Ethenoadenosine
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cell Biology
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2 λ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.